4-Nitrothiobenzamid 97

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H52140.03 H52140.06
H52140.03EA 894 DKK
H52140.03 H52140.06
4-Nitrothiobenzamid 97
4-Nitrothiobenzamid

Purity: 97% CAS No.: 26060-30-0 Molecular Formula: C7H6N2O2S Formula Weight: 182.20 MDL No.: MFCD06150000

Formel: C₇H₆N₂O₂S
Molekylvægt: 182.2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06150000
CAS-nummer: 26060-30-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR