4-Hydroxycarbazol 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 9H-Carbazol-4-ol, Carbazol-2-ol

366960010 366960050
ACRO366960010EA 327 DKK
ACRO366960010 ACRO366960050
4-Hydroxycarbazol 98+%
4-Hydroxycarbazole
Formel: C₁₂H₉NO
Smeltepunkt: 169…173 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02178385
CAS-nummer: 52602-39-8
EINECS: 258-034-9

Bestil nu

Specifikationer Testresultater

Appearance Tan to dark brown Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR