4-Bromimidazol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

376900010 376900050
ACRO376900010EA 617 DKK
ACRO376900010 ACRO376900050
4-Bromimidazol 97%
4-Bromoimidazole
Formel: C₃H₃BrN₂
Smeltepunkt: 126…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00047021
CAS-nummer: 2302-25-2
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to beige Powder or flakes
Infrared spectrum Conforms
Melting point 126 to 132 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR