Genistein (synthetic) 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

328270250 328271000
ACRO328270250EA 1190 DKK
ACRO328270250 ACRO328271000
Genistein (synthetic) 99%
Genistein
Formel: C₁₅H₁₀O₅
Smeltepunkt: 297…298 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00016952
CAS-nummer: 446-72-0
EINECS: 207-174-9

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR