2-Furonitril 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Cyanofuran
170430050
170430250
ACRO170430050EA
621
DKK
ACRO170430050
ACRO170430250
2-Furonitril 99%
2-Furonitril
Formel:
C₅H₃NO Molekylvægt: 93.08 g/mol Kogepunkt: 146 Massefylde: 1.06 g/cm³ Flammepunkt: 35 °C |
MDL Number:
MFCD00003223 CAS-nummer: 617-90-3 EINECS: 210-537-4 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Clear light yellow to light orange Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4780 to 1.4800 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...