5-Nitro-2-furonitril ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 5-Nitrofuran-2-carbonitrile, 2-Cyano-5-nitrofuran

L10486.06 L10486.14
L10486.06EA 1030 DKK
L10486.06 L10486.14
5-Nitro-2-furonitril ≥97%
5-Nitro-2-furonitril

Purity: 97% CAS No.: 59-82-5 Molecular Formula: C5H2N2O3 Formula Weight: 138.08 Melting Point: 61-64° Beilstein Reference: 123933 UN No.: 3439 MDL No.: MFCD00082647

Formel: C₅H₂N₂O₃
Molekylvægt: 138.08 g/mol
Smeltepunkt: 61…64 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00082647
CAS-nummer: 59-82-5
UN nummer: 3439
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR