1-Acetyl-2-naphthol ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Acetyl-2-hydroxynaphthalene, 2-Hydroxy-1-acetonaphthone, 1-(2-Hydroxynaphthalen-1-yl)ethanone, 2'-Hydroxy-1'-acetonaphthone, 1-(2-Hydroxy-1-naphthyl)ethan-1-one

A18537.06 A18537.14
A18537.06EA 358 DKK
A18537.06 A18537.14
1-Acetyl-2-naphthol ≥99%
1-Acetyl-2-naphthol

Purity: 99% CAS No.: 574-19-6 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 63-67° Boiling Point: 130-132°/2mm MDL No.: MFCD00012131

Formel: C₁₂H₁₀O₂
Kogepunkt: 130…132 °C (2 mmHg)
Smeltepunkt: 63…67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012131
CAS-nummer: 574-19-6
EINECS: 209-369-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR