3-Brom-5-chlorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Bromo-5-chlorobenzonitile

B3118-1G B3118-5G
TCIAB3118-1GEA 279 DKK
TCIAB3118-1G TCIAB3118-5G
3-Brom-5-chlorbenzonitril ≥98.0% (by GC)
3-Brom-5-chlorbenzonitril
Formel: C₇H₃BrClN
Kogepunkt: 144°C
MDL Number: MFCD06200840
CAS-nummer: 304854-55-5
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=908a91176898e1bd9a9a01f984eaa4d694edb55b&vid=908a91176898e1bd9a9a01f984eaa4d694edb55b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR