4-(Dimethylamino)benzophenon 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B20389.06 B20389.14 B20389.22
B20389.06EA 739 DKK
B20389.06 B20389.14 B20389.22
4-(Dimethylamino)benzophenon 98%
4-(Dimethylamino)benzophenon

Purity: 98% CAS No.: 530-44-9 Molecular Formula: C15H15NO Formula Weight: 225.29 Melting Point: 88-90° Merck Reference: 14,3233 MDL No.: MFCD00008311

Formel: C₁₅H₁₅NO
Kogepunkt: 245…250 °C (15 mmHg)
Smeltepunkt: 88…90 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008311
CAS-nummer: 530-44-9
EINECS: 208-478-4
Merck Index: 14,3233

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR