Pasteur pipettes, disposable
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
747750 747755 747760 747765 747770 747775
BRND747750EA 580 DKK
BRND747750 612-1714 BRND747760 BRND747765 612-1715 BRND747775
Pasteur pipettes, disposable
Pipetter Pasteur-/Overførselspipetter
LDPE.
  • Excellent at reproducing number of drops per mililitre
  • Ideal for aliquots
  • Pasteur pipettes can be filled and deep frozen, or changed into a closed vessel by heat-sealing the tip
  • The integrated pipetting bulb depresses easily, minimising fatigue from frequent pipetting
  • Resistant to gas or gamma radiation sterilisation