Reading pocket magnifiers, mobilux
Leverandør: ESCHENBACH OPTIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15117 15113 151112 15112 15116 15114 15122
630-1313EA 1250 DKK
630-1313 630-1312 630-2178 630-2179 630-2180 620-2616 ESCH15122
Reading pocket magnifiers, mobilux
Forstørrelsesglas og lupper
LED illumination and coated lens on both sides with Ceratec® anti-scratch coating for optimised performances.
  • Exchangeable white light LED
  • Attachable yellow filter of 50000 hours
  • Easy handling and large light switch
Leveringsinformation: Supplied with two AA batteries and case.