Control seals, close-it food & pharma
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5303-2017 5303-2018 5303-3018 5303-3017 5303-2020 5303-2023 5303-2021 5303-2022 5303-3020 5303-3022 5303-3021 5303-3023 9990-8075
312-0073EA 1270 DKK
312-0073 312-0074 312-0076 312-0075 312-0094 312-0097 312-0095 312-0096 312-0098 312-0100 312-0099 312-0101 BURK9990-8075
Control seals, close-it food & pharma
Etiketter
A control seal with an adhesive approved for use with foodstuffs. The strength of the adhesive is almost as powerful as that of the original. Just like the original, close-it food and pharma has a barrier layer of aluminium foil.
  • Adheres very firmly
  • Can be written on
  • Adhesive complies with the FDA and European guidelines for foodstuffs
Suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods where cleanliness is particularly important.