Hexanal ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Capronaldehyde

A16265.22 A16265.36
A16265.22EA 485 DKK
A16265.22 A16265.36
Hexanal ≥98%
Hexanal

Purity: 98% CAS No.: 66-25-1 Molecular Formula: CH3(CH2)4CHO Formula Weight: 100.16 Flash Point: 25°(77°F) Refractive Index: 1.4050 Melting Point: -56° Boiling Point: 130-131° Beilstein Reference: 506198 Merck Reference: 14,1760 UN No.: 1207 MDL No.: MFCD00007027

Formel: C₆H₁₂O
Molekylvægt: 100.16 g/mol
Kogepunkt: 130…131 °C
Smeltepunkt: –56 °C
Massefylde: 0.834
Flammepunkt: 25 °C (77 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007027
CAS-nummer: 66-25-1
EINECS: 200-624-5
UN nummer: 1207
ADR: 3 III
Merck Index: 13,01766

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR