Hexanal 96% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Capronaldehyde

151850010 151855000 151850025
ACRO151850010EA 1910 DKK
ACRO151850010 ACRO151855000 ACRO151850025
Hexanal 96% stabilised
Hexanal
Formel: C₆H₁₂O
Kogepunkt: 130…131 °C (760 mmHg)
Smeltepunkt: –56 °C
Massefylde: 0,815 g/cm³
Flammepunkt: 32 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007027
CAS-nummer: 66-25-1
EINECS: 200-624-5
UN nummer: 1207
ADR: 3,III
Merck Index: 13,01766

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4070 (20°C, 589 nm)
Density (20°C) 0.810 to 0.820 g/ml
GC ≥95 %
Stabilizer 100 to 500 ppm alpha-Tocopherol
Acid value ≤10 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR