Butyraldehyd 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Butanal
108090010
108091000
108090025
ACRO108090010EA
411
DKK
ACRO108090010
ACRO108091000
ACRO108090025
Butyraldehyd 99%
Butyraldehyd
Formel:
C₄H₈O Kogepunkt: 75 °C (760 mmHg) Smeltepunkt: –96 °C Massefylde: 0,817 g/cm³ Flammepunkt: –12 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007023 CAS-nummer: 123-72-8 EINECS: 204-646-6 UN nummer: 1129 ADR: 3,II Merck Index: 13,01591 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3770 to 1.3830 (20°C, 589 nm) |
GC | ≥98.5 % |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...