2-Brom-2-nitropropan ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A18385.06 A18385.14
A18385.06EA 374 DKK
A18385.06 A18385.14
2-Brom-2-nitropropan ≥98%
2-Bromo-2-nitropropane

Purity: 98% CAS No.: 5447-97-2 Molecular Formula: C3H6BrNO2 Formula Weight: 168.00 Flash Point: 61°(141°F) Refractive Index: 1.4640 Boiling Point: 45-46°/16mm Beilstein Reference: 1746626 MDL No.: MFCD00007389

Formel: H₃CBrC(NO₂)CH₃
Molekylvægt: 167.99 g/mol
Kogepunkt: 45…46 °C (16 mmHg)
Smeltepunkt: 33…35 °C
Massefylde: 1.584
Flammepunkt: 61 °C (141 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007389
CAS-nummer: 5447-97-2
EINECS: 226-668-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR