1-Brom-2-chlorethan 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
214181000
214185000
ACRO214181000EA
399
DKK
ACRO214181000
ACRO214185000
1-Brom-2-chlorethan 98%
1-Bromo-2-chloroethane
Formel:
BrH₂CCH₂Cl Kogepunkt: 106…107 °C (760 mmHg) Smeltepunkt: –17 °C Massefylde: 1,73 g/cm³ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000962 CAS-nummer: 107-04-0 EINECS: 203-456-0 UN nummer: 2810 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4900 to 1.4920 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...