1,2-Dibromtetrachlorethan 97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
B20276.14 B20276.22
B20276.14EA 428 DKK
B20276.14 B20276.22
1,2-Dibromtetrachlorethan 97%
1,2-Dibromotetrachloroethane
Purity: 97% CAS No.: 630-25-1 Molecular Formula: BrCCl2CCl2Br Formula Weight: 325.65 Melting Point: ca 220° dec. Beilstein Reference: 1699471
Formel: BrCl₂CCBrCl₂
Kogepunkt: 222 °C (1013 hPa)
Smeltepunkt: ∼220 °C (dec.)
Massefylde: 2,71
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000780
CAS-nummer: 630-25-1
EINECS: 211-136-7