1-Bromdodecan ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A10600.0B A10600.0I A10600.30
A10600.0BEA 1400 DKK
A10600.0B A10600.0I A10600.30
1-Bromdodecan ≥98%
1-Bromododecane

Purity: 98% CAS No.: 143-15-7 Molecular Formula: C12H25Br Formula Weight: 249.24 Flash Point: 146°(294°F) Refractive Index: 1.4580 Odour: Weak Melting Point: -10° Boiling Point: 276° Beilstein Reference: 506159 Merck Reference: 14,5389 MDL No.: MFCD00000225

Formel: BrH₂C(CH₂)₁₀CH₃
Kogepunkt: 276 °C
Smeltepunkt: –10 °C
Massefylde: 1,040
Flammepunkt: 146 °C (294 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000225
CAS-nummer: 143-15-7
EINECS: 205-587-9
Merck Index: 14,5389

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR