1-Bromdodecan 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
106910010
106912500
106910025
ACRO106910010EA
991
DKK
ACRO106910010
ACRO106912500
ACRO106910025
1-Bromdodecan 98%
1-Bromododecane
Formel:
BrH₂C(CH₂)₁₀CH₃ Kogepunkt: 134…135 °C (6 mmHg) Smeltepunkt: 11…9 °C Massefylde: 1,03 g/cm³ Flammepunkt: 110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000225 CAS-nummer: 143-15-7 EINECS: 205-587-9 Merck Index: 13,05406 |
Specifikationer Testresultater
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4590 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥97.5 % |
Color scale | ≤125 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...