1,2-Dibromethan ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 99%
CAS No.: 106-93-4
Molecular Formula: C2H4Br2
Formula Weight: 187.87
Flash Point: None
Refractive Index: 1.5385
Melting Point: 9-10°
Boiling Point: 132°
Merck Reference: 14,3796
UN No.: 1605
MDL No.: MFCD00000233
Formel:
BrH₂CCH₂Br Kogepunkt: 132 °C Smeltepunkt: 9…10 °C Massefylde: 2,179 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000233 CAS-nummer: 106-93-4 EINECS: 203-444-5 UN nummer: 1605 ADR: 6.1,I Merck Index: 14,3796 |