Neodymiumtrinitrat hexahydrat ≥99.9%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Neodymium (III) nitrate hexahydrate
12912.18
12912.30
12912.18EA
1060
DKK
12912.18
12912.30
Neodymiumtrinitrat hexahydrat ≥99.9%
Neodymiumtrinitrat hexahydrat
Purity: 99.9%
CAS No.: 16454-60-7
Molecular Formula: Nd(NO3)3.6H2O
Formula Weight: 438.34 (330.25anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
pH: 3.0-4.0
Melting Point: ca 40°
Merck Reference: 14,6450
UN No.: 1477
MDL No.: MFCD00149799
Formel:
N₃NdO₉·6H₂O Smeltepunkt: ∼40 °C Massefylde: 2,265 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149799 CAS-nummer: 16454-60-7 EINECS: 233-153-9 UN nummer: 1477 ADR: 5.1,III Merck Index: 14,6450 |