Tetrahydrofuran ≥99.7% stabilised for HPLC, non UV
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Butylene oxide, THF, Diethylene oxide
Stabilization:
stabiliseret med BHT/Ionol 250 ppm
Stabilized with 250 ppm BHT
Purity: 99.7+%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Formel:
C₄H₈O Kogepunkt: 66 °C Smeltepunkt: –108 °C Massefylde: 0,889 Flammepunkt: –17 °C (1 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005356 CAS-nummer: 109-99-9 EINECS: 203-726-8 UN nummer: 2056 ADR: 3,II REACH: 05-2114615188-48 Merck Index: 14,9211 |