1-Brom-2-iodbenzen ≥98% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene

A17020.06 A17020.14 A17020.22
A17020.06EA 425 DKK
A17020.06 A17020.14 A17020.22
1-Brom-2-iodbenzen ≥98% stabilised
1-Brom-2-iodbenzen

Stabilized with copper


Purity: 98+% CAS No.: 583-55-1 Molecular Formula: C6H4BrI Formula Weight: 282.91 Flash Point: 110°(230°F) Refractive Index: 1.6645 Melting Point: 9-10° Boiling Point: 257° Beilstein Reference: 1855309 MDL No.: MFCD00001030

Formel: C₆H₄BrI
Molekylvægt: 282.91 g/mol
Kogepunkt: 257 °C
Smeltepunkt: 9…10 °C
Massefylde: 2.256
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001030
CAS-nummer: 583-55-1
EINECS: 209-508-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR