Natriumkarbonat decahydrat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Soda

K303.1 K303.2
ROTHK303.1EA 708.34 DKK
ROTHK303.1 ROTHK303.2
Natriumkarbonat decahydrat
Natriumkarbonat decahydrat
Formel: Na₂CO₃·10H₂O
Smeltepunkt: 33 °C
Massefylde: 1,46 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149178
CAS-nummer: 6132-02-1
EINECS: 207-838-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR