Natriumthiosulfat 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Leverandør: REAGECON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
T20105 T20101
REGET20105EA 771 DKK
REGET20105 REGET20101
Natriumthiosulfat 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Sodium thiosulphate
Formel: Na₂S₂O₃
Molekylvægt: 158.11 g/mol
Kogepunkt: 100
Smeltepunkt: 48
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003499
CAS-nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844