Natriumthiosulfat 0.1 M Na2S2O3 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Leverandør: Honeywell Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
35245-1L
35245-10L-VP
35245-5L-VP
35245-6X1L
35245-1LEA
383
DKK
35245-1L
35245-10L-VP
35245-5L-VP
35245-6X1L
Natriumthiosulfat 0.1 M Na2S2O3 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Sodium thiosulphate
Formel:
Na₂S₂O₃ Molekylvægt: 158.11 g/mol Kogepunkt: 100 °C (1.013 hPa) Smeltepunkt: 48 Massefylde: 1.0 g/cm³ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003499 CAS-nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Specifikationer Testresultater
Factor | 0.999 - 1.001 |