Hot plates, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6796-400D 6796-600D 6798-400D 6798-600D
444-1006EA 4080 DKK
444-1006 444-1009 444-1012 444-1015
Hot plates, Corning®
Varmeapparater Varmeplader
Corning® hotplates with Pyroceram® glass-ceramic top, rugged housing and heating elements and digital temperature display.
  • Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Optional external temperature controller (444-1004) truly controls temperature inside the vessel
  • Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned off
  • Two piece casting design deflects spills away from electronics and includes a built in mount for an optional 457 mm support rod
  • Two year manufacturer's warranty
Separate temperature sensors will provide power cut-off if the unit overheats. All units have a small footprint and low profile to ensure a comfortable fit inside hoods or storage cabinets. The extremely durable and heat-resistant Pyroceram® tops provide uniform heating, are easy-to-clean, and are highly resistant to scratches, corrosion, and chemicals. The angled front panel and large knob design minimises damage resulting from spills. The optional external temperature controller ensures precision temperature accuracy of the liquid inside the vessel.
Certificeringer: UL and C-UL listed.