Screw caps for media storage bottles, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1395-45LTMC 1395-25TS 1395-32HTC 1395-32LTC 1395-32LTR 1395-32TS 1395-45HTC 1395-45HTR 1395-45HTSC 1395-45LTC 1395-45LTC1 1395-45LTR 1395-45SS 1395-45TS
734-2288EA 515 DKK
734-2288 CORN1395-25TS CORN1395-32HTC CORN1395-32LTC CORN1395-32LTR CORN1395-32TS CORN1395-45HTC CORN1395-45HTR CORN1395-45HTSC 734-2287 CORN1395-45LTC1 CORN1395-45LTR CORN1395-45SS CORN1395-45TS
Screw caps for media storage bottles, Corning®
Låg Flaskelåg
Screw caps and pouring rings, made of PP or PBT.
  • For laboratory/media bottles
  • Plug seal cap ideal for media preparation work
  • Autoclavable up to 121 °C (PP) and dry heat sterilized at up to 180 °C PBT
Open top caps are designed to be used with piercible silicone septa (sold separately). Close top caps made of PP with linerless plug seal and close top caps made of PBT with PTFE liner for efficient sealing.