Natriumhydroxid standard solution 0.2 mol/l (0.2 N) in water
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Caustic soda
349685000 349680025
ACRO349685000EA 230 DKK
ACRO349685000 ACRO349680025
Natriumhydroxid standard solution 0.2 mol/l (0.2 N) in water
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: ∼105 °C (1013 hPa)
Smeltepunkt: 323
Massefylde: 1.01 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
Merck Index: 14,08627
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless Solution
Strength 0.1996 to 0.2004 N