Stopcocks, straight ot T-bore type
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88610 88615 88620 88625 88630 88635 88640 88650 88655 88660 88665 88670 88675 88680
BRND88610EA 436 DKK
BRND88610 BRND88615 BRND88620 BRND88625 BRND88630 BRND88635 BRND88640 BRND88650 BRND88655 227-1115 BRND88665 BRND88670 BRND88675 BRND88680
Stopcocks, straight ot T-bore type
Stophaner
PTFE, with PP valve lever.
  • Excellent chemical resistance
  • Max. pressure 1 bar
  • Can be sterilised at 135 °C
With two or three integrated hose nozzles.