Slangekonnektorer, Y-formede
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
152900 152903 152905 152907 152910
BRND152900EA 216 DKK
BRND152900 BRND152903 BRND152905 BRND152907 BRND152910
Slangekonnektorer, Y-formede
Fittings og stik
PP, translucent.
  • Good mechanical strength and chemical resistance
  • Excellent thermal resistance
  • Autoclavable
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine 'grip'. This will then give equivalent crest dimension for the particular connector concerned.