Cryoboxes for upright/chest freezers, HERAfreeze®
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50058604 50058908 50058911 50058914 50058916 50058917 50058918 50058919 50059050 50059063 50059064 50059067
HERA50058604EA 112 DKK
HERA50058604 HERA50058908 HERA50058911 HERA50058914 HERA50058916 HERA50058917 HERA50058918 HERA50058919 390-4400 390-4408 390-4409 390-4411
Cryoboxes for upright/chest freezers, HERAfreeze®
Bokse Æsker til Kryorør
Cryoboxes for upright and chest freezer.
Fiberboard
Light, flexible and economical.
Polycarbonate
Robust, flexible, and temperature resistant.
Polypropylene
Robust and autoclavable.