Kaliumchlorid 3 M aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Potassiumchloride
43398.K2 43398.AK
43398.K2EA 1240 DKK
43398.K2 43398.AK
Kaliumchlorid 3 M aqueous solution
Potassium chloride
CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00011360
For refilling calomel electrodes
Formel: KCl
Molekylvægt: 74.55 g/mol
Kogepunkt: 1420
Smeltepunkt: 773
Massefylde: 1.13 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621