Osmiumtetraoxid 4% in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Osmiumsyre
45385.AB 45385.9W
45385.ABEA 2240 DKK
45385.AB 45385.9W
Osmiumtetraoxid 4% in aqueous solution
Osmiumsyre
CAS No.: 20816-12-0 Molecular Formula: OsO4 Formula Weight: 254.10 Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Chlorine-like Merck Reference: 14,6893 UN No.: 3287 MDL No.: MFCD00011150
Formel: O₄Os
Molekylvægt: 254.23 g/mol
Kogepunkt: 130
Smeltepunkt: 40
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011150
CAS-nummer: 20816-12-0
EINECS: 244-058-7
UN nummer: 3287
ADR: 6.1 III
Merck Index: 14,6893