Universalkar, med dækkener, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DS5300-0507 DS5300-9609 DS5300-9910
216-8262EA 854 DKK
216-8262 216-8264 216-8265
Universalkar, med dækkener, Nalgene®
Dåser Dåser, allround
Polycarbonate, clear.
  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.