Membranfiltre, MembraClear, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7141-025 7141-047
514-0382EA 1660 DKK
514-0382 514-0383
Membranfiltre, MembraClear, Whatman™
Filtre Membranfiltre
These mixed cellulose ester membrane filters are designed for asbestos sampling using the membrane filter method for phase contrast microscopy. Asbestos sampling isolates these fibres from circulating air to determine concentrations.
  • Designed for use with the 'hot block method'
  • White with black grid for high contrast during asbestos testing using microscopy
  • Becomes transparent with acetone vapour treatment, distinguishing fibres from crystalline artifacts