ortho-Phosphorsyre ≥85% (w/w) in aqueous solution ACS
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Phosphorsyre, Fosforsyre
33266.22 33266.A1 33266.D6
33266.22EA 521 DKK
33266.22 33266.A1 33266.D6
ortho-Phosphorsyre ≥85% (w/w) in aqueous solution ACS
ortho-Phosphorsyre
CAS No.: 7664-38-2 Molecular Formula: H3O4P Formula Weight: 98.00 Physical Form: Viscous liquid Density: 1.70 Refractive Index: 1.4310 Melting Point: 19-21° Merck Reference: 14,7344 UN No.: 1805 MDL No.: MFCD00011340
Formel: H₃PO₄
Molekylvægt: 98 g/mol
Kogepunkt: 158
Smeltepunkt: 19…21 °C
Massefylde: 1.70
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011340
CAS-nummer: 7664-38-2
EINECS: 231-633-2
UN nummer: 1805
ADR: 8 III
Merck Index: 14,7344