Connection system GL 45, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1129750 1129751
554-3000EA 601 DKK
554-3000 554-3001
Connection system GL 45, DURAN®
Låg Flaskelåg
Connection system with PP screw cap GL 45 with two or three PTFE ports. Flexible modular system. Typical applications: safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
  • Four tubing sizes (1,6, 3,0, 3,2 or 6,0 mm) can be used by selecting the correct insert
  • Sterile pressure equalisation is possible through use of membrane filter
  • Unused ports can be provided with a blind cap