Microcentrifuge tubes, 0,65/1,7/2,0 ml, with snap cap, Costar®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3207 3208 3209 3213 3620 3621 3622
525-0008EA 299 DKK
525-0008 525-0009 525-0010 525-0011 525-0012 525-0013 525-0014
Microcentrifuge tubes, 0,65/1,7/2,0 ml, with snap cap, Costar®
Rør Mikrocentrifugerør
PP, graduated, autoclavable.
  • External graduations and frosted writing spot for easy sample identification
  • Positive seal design allows for repeated opening and closing
  • Flat cap surface for convenient labelling
  • Withstands a maximum RCF of 17000×g
These microcentrifuge tubes are made of natural (uncoloured) or coloured polypropylene and are certified RNase-/DNase-free.
Assorted colour pack includes one bag each of blue, green, yellow, red and orange tubes.
Bestillingsoplysninger: Optional autoclavable plastic tube racks sold separately.