VWR®, Measuring cylinders, low form, class B, borosilicate glass
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-3846EA 496 DKK
612-3846 612-3845 612-3844 612-3843 612-3842 612-3841 612-3847
VWR®, Measuring cylinders, low form, class B, borosilicate glass
Cylindere Målecylindre
Borosilicate 3.3 glass, class B, low form.
  • Hexagonal glass base, with spout
  • High contrast amber stain graduations with ring marks at major graduations
DIN 12680
Certificeringer: DIN 12680