Cell culture dishes, non treated, for suspension culture, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
171099 150340
734-2109EA 4120 DKK
734-2109 734-2374
Cell culture dishes, non treated, for suspension culture, Nunc™
Dyrkningsplader Celledyrkningsskåle
PS, with lid, sterile. Non treated for suspension culture. Optically clear and suitable for microscopy.