Membranskivefiltre til ultrafiltrering, Omega™
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OM001043 OM001076 OM005043 OM005076 OM010043 OM010062 OM010076 OM030043 OM030062 OM030076 OM050076 OM100043 OM100076
516-8571EA 3040 DKK
516-8571 516-8574 516-8589 516-8592 516-8607 516-8609 516-8610 516-8616 516-8618 516-8619 516-8628 516-8643 516-8646
Membranskivefiltre til ultrafiltrering, Omega™
Filtre Membranfiltre
Replacement membranes for other stirred cell systems. Used for concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules. Discs can be washed and reused.
Omega™ membrane, low protein binding modified PES.
pH range: 1 to 14