Nalgene® Media Bottles, Square, with Screw Cap
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
322020-0125 322020-0030 322020-0060 322020-0250 322020-0500 322020-1000 322020-2000
NALG322020-0125EA 2450 DKK
NALG322020-0125 NALG322020-0030 216-2987 NALG322020-0250 NALG322020-0500 NALG322020-1000 NALG322020-2000
Nalgene® Media Bottles, Square, with Screw Cap
Flasker Medieflasker
PETG bottle with HDPE screw closure.
  • Graduated
  • Durable
  • Break-resistant
  • Leakproof
Package and ship liquid media, buffers and sera using square media bottles with screw caps. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera. Non-sterile bottle offers excellent gas barrier properties; clear PETG material allows for easy identification of contents.
Certificeringer: USP Class VI, FDA Compliance 21 CFR