Erlenmeyer-kolber, smal eller bred hals, med krave
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
92773 92717 92724 92736 92739 92744 92754 92763 92768 92814 92817 92824 92836 92839 92844 92854 92863
BRND92773EA 1140 DKK
BRND92773 BRND92717 BRND92724 BRND92736 BRND92739 BRND92744 BRND92754 BRND92763 BRND92768 BRND92814 BRND92817 BRND92824 BRND92836 BRND92839 BRND92844 BRND92854 BRND92863
Erlenmeyer-kolber, smal eller bred hals, med krave
Kolber Erlenmeyer-kolber
Borosilicate glass, grade 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.
Certificeringer: ISO 1773 narrow mouth (except 200 and 300 ml)
DIN ISO 24450 wide mouth (except 25, 200, 300 and 2000 ml).