Natriumdodecylsulfat (SDS) 200 g/l in aqueous solution
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
20767.01
20767.02
20767.03
SERA20767.01EA
179.14
DKK
SERA20767.01
SERA20767.02
SERA20767.03
Natriumdodecylsulfat (SDS) 200 g/l in aqueous solution
Natriumdodecylsulfat (SDS)
Formel:
CH₃(CH₂)₁₁OSO₃Na Massefylde: 1…1,1 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00036175 CAS-nummer: 151-21-3 EINECS: 205-788-1 |