Flasker, bred hals, HDPE, emballage, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312184-0004 312184-0008 312184-0016
NALG312184-0004EA 6230 DKK
NALG312184-0004 NALG312184-0008 NALG312184-0016
Flasker, bred hals, HDPE, emballage, med skruelåg, Nalgene®
Flasker Plastflasker
HDPE, amber translucent, with opaque amber PP screw cap.
  • Protects light sensitive products
  • Leakproof
  • Good chemical and temperature resistance
Wide mouth allows easy filling and dispensing of liquids and powders.
Meets the requirements of light-resistant containers per USP latest edition.