Natriumdodecylsulfat (SDS) 100 g/l in aqueous solution, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ100
1.06022.1000 1.06022.9025
1.06022.1000EA 1170 DKK
1.06022.1000 1.06022.9025
Natriumdodecylsulfat (SDS) 100 g/l in aqueous solution, Sigma-Aldrich®
Natriumdodecylsulfat (SDS)
Formel: CH₃(CH₂)₁₁OSO₃Na
Molekylvægt: 288.38 g/mol
Massefylde: 1…1.1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036175
CAS-nummer: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM