Formaldehyd 37% in aqueous solution
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
7398.1 7398.2 7398.3
ROTH7398.1EA 0 DKK
ROTH7398.1 ROTH7398.2 ROTH7398.3
Formaldehyd 37% in aqueous solution
Formaldehyd
Formel: CH₂O
Molekylvægt: 30.03 g/mol
Kogepunkt: -19.3
Smeltepunkt: -92
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8
UN nummer: 2209
ADR: 8 III
Merck Index: 13,04259