Bægre, lav type, ETFE
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
90224 90228 90238 90248 90252 90254 90256 90262
BRND90224EA 132 DKK
BRND90224 BRND90228 BRND90238 BRND90248 BRND90252 BRND90254 BRND90256 BRND90262
Bægre, lav type, ETFE
Bægre
ETFE, translucent.
  • Excellent chemical resistance
  • Good thermal resistance from –100 to +150 °C
  • Autoclavable
With spout and easy to read blue printed graduations.
Certificeringer: ISO 7056